Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tryptamine, 98+%

Catalog No. p-7081230
Click to view available options
Quantity:
50 g

CAS: 61-54-1 | C10H12N2 | 160.22 g/mol

Tryptamine is commonly used in the preparation of biologically active compounds such as neurotransmitters and psychedelics, multi-layered analysis, used to differentiate and identify biogenic amines1

  • in the synthesis of indole ring-terminated thiol, ω-mercaptooctyltryptamide.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Tryptamine is commonly used in the preparation of biologically active compounds such as neurotransmitters and psychedelics, multi-layered analysis, used to differentiate and identify biogenic amines1 • in the synthesis of indole ring-terminated thiol, ω-mercaptooctyltryptamide.

Solubility
Soluble in DMSO, Methanol. Insoluble in water.

Notes
Store in cool, dry conditions in well sealed containers. Keep container tightly closed.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 61-54-1
Molecular Formula C10H12N2
Molecular Weight (g/mol) 160.22
MDL Number MFCD00005661
InChI Key APJYDQYYACXCRM-UHFFFAOYSA-N
Synonym tryptamine, 2-1h-indol-3-yl ethanamine, 3-2-aminoethyl indole, 1h-indole-3-ethanamine, 2-3-indolyl ethylamine, indol-3-ethylamine, indole, 3-2-aminoethyl, 2-indol-3-yl ethylamine, 2-1h-indol-3-yl ethan-1-amine, tryptamin
PubChem CID 1150
ChEBI CHEBI:16765
IUPAC Name 2-(1H-indol-3-yl)ethanamine
SMILES NCCC1=CNC2=CC=CC=C12

Specifications

Melting Point 111°C to 116°C
Boiling Point 137°C (0.15 mmHg)
Flash Point 185°C (365°F)
Quantity 50 g
Beilstein 125513
Sensitivity Air sensitive
Merck Index 14,9796
Solubility Information Soluble in DMSO,Methanol. Insoluble in water.
Formula Weight 160.22
Percent Purity ≥98%
Chemical Name or Material Tryptamine
Show More Show Less
Hazard Category Warning
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
EINECSNumber 200-510-5
RTECSNumber NL4020000
TSCA No
Recommended Storage Keep cold; Store under Argon

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.