Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Tryptase alpha/beta 1 Antibody (RM0355-10H34), Alexa Fluor™ 350, Novus Biologicals™

Rat Monoclonal Antibody
Supplier: Novus Biologicals NBP211983AF350
Description
Tryptase alpha/beta 1 Monoclonal antibody specifically detects Tryptase alpha/beta 1 in Mouse samples. It is validated for Western Blot.Specifications
Tryptase alpha/beta 1 | |
Monoclonal | |
Alexa Fluor 350 | |
50 mM sodium borate with 0.05% sodium azide | |
EC:3.4.21.59, epididymis secretory sperm binding protein, mast cell alpha II tryptase, mast cell beta I tryptase, mast cell tryptase, TPS1, TPS2, TPSB1, TPSB2, tryptase alpha II, Tryptase alpha-1, tryptase beta I, tryptase beta-1, Tryptase beta-2, Tryptase I, tryptase II, tryptase-1, tryptase-2, tryptase-I, tryptase-III | |
Rat | |
Protein A or G purified | |
Immunology, metabolism | |
7177 | |
Store at 4°C in the dark. |
Western Blot | |
RM0355-10H34 | |
Western Blot | |
Q02844 | |
TPSAB1 | |
Mouse recombinant Tryptase alpha/beta 1 | |
0.1 mL | |
Primary | |
Mouse | |
IgG2 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction