Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Tryptase alpha/beta 1 Antibody (SR1510), Novus Biologicals™

Rabbit Monoclonal Antibody
$423.00
Specifications
Antigen | Tryptase alpha/beta 1 |
---|---|
Clone | SR1510 |
Dilution | Western Blot 1:500-1:2000, Immunohistochemistry 1:50-1:200, Immunoprecipitation 1:50 |
Applications | Western Blot, Immunohistochemistry, Immunoprecipitation |
Classification | Monoclonal |
Description
Tryptase alpha/beta 1 Monoclonal antibody specifically detects Tryptase alpha/beta 1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, ImmunoprecipitationSpecifications
Tryptase alpha/beta 1 | |
Western Blot 1:500-1:2000, Immunohistochemistry 1:50-1:200, Immunoprecipitation 1:50 | |
Monoclonal | |
Purified | |
RUO | |
Human, Mouse, Rat | |
EC:3.4.21.59, epididymis secretory sperm binding protein, mast cell alpha II tryptase, mast cell beta I tryptase, mast cell tryptase, TPS1, TPS2, TPSB1, TPSB2, tryptase alpha II, Tryptase alpha-1, tryptase beta I, tryptase beta-1, Tryptase beta-2, Tryptase I, tryptase II, tryptase-1, tryptase-2, tryptase-I, tryptase-III | |
A synthesized peptide derived from human Tryptase alpha/beta 1 (Uniprot #: Q15661) | |
Primary | |
Store at -20° C. Avoid freeze/thaw cycles. |
SR1510 | |
Western Blot, Immunohistochemistry, Immunoprecipitation | |
Unconjugated | |
Rabbit | |
Immunology, metabolism | |
PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
7177 | |
IgG | |
Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title