Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Tryptase alpha/beta 1 Antibody (TPSAB1/1961) - Azide and BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP275754
Description
Tryptase alpha/beta 1 Monoclonal antibody specifically detects Tryptase alpha/beta 1 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin), Protein Array.Specifications
Tryptase alpha/beta 1 | |
Monoclonal | |
0.2 mg/ml | |
Immunohistochemistry-Paraffin 1-2 ug/ml | |
Q15661 | |
TPSAB1 | |
Recombinant human Tryptase alpha/TPS1 protein fragment (around aa 115-233) (Exact sequence is proprietary) (Uniprot: Q15661) | |
100 μg | |
Primary | |
Human | |
Purified |
Immunohistochemistry (Paraffin) | |
TPSAB1/1961 | |
Unconjugated | |
10 mM PBS with No Preservative | |
EC:3.4.21.59, epididymis secretory sperm binding protein, mast cell alpha II tryptase, mast cell beta I tryptase, mast cell tryptase, TPS1, TPS2, TPSB1, TPSB2, tryptase alpha II, Tryptase alpha-1, tryptase beta I, tryptase beta-1, Tryptase beta-2, Tryptase I, tryptase II, tryptase-1, tryptase-2, tryptase-I, tryptase-III | |
Mouse | |
Protein A or G purified | |
Immunology, metabolism | |
This antibody reacts with mast cells distributed in skin, synovium, lung, and heart. This antibody does not bind with any other cell type. Human mast cell tryptase is considered to be an important marker of mast cell activation and is an important mediator of inflammation. Mastocytosis is a term collectively used for a group of disorders in which there is abnormal accumulation of mast cells in one or multiple organs. Anti-tryptase, combined with anti-CD2, anti-CD25, and anti-CD117, can be useful in identifying reactive mast cell hyperplasia, myelogenous neoplasms, mast cell leukemia, and mastocytosis. Due to sequence identity this antibody will likely cross react with tryptase beta 2. | |
Store at 4°C. | |
IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction