Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tryptase alpha/beta 1 Antibody (RM0355-10H34) - Azide and BSA Free, Novus Biologicals™
SDP

Catalog No. p-7200469 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB018977 0.025 mg
NBP211983 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB018977 Supplier Novus Biologicals Supplier No. NBP2119830.025MG
Only null left
Add to Cart
Add to Cart

Rat Monoclonal Antibody

Tryptase alpha/TPS1 Monoclonal specifically detects Tryptase alpha/TPS1 in Mouse samples. It is validated for Western Blot.

Specifications

Antigen Tryptase alpha/TPS1
Applications Western Blot
Classification Monoclonal
Clone RM0355-10H34
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1:250-1000
Formulation A 0.2 um filtered solution in PBS with No Preservative
Gene Accession No. Q02844
Gene Alias TPS1, TPS2, TPSAB1, TPSB1, tryptase alpha/beta 1
Gene Symbols TPSAB1
Host Species Rat
Immunogen Mouse TPSAB1
Purification Method Protein G purified
Quantity 0.025 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7177
Reconstitution Reconstitute with 0.2 ml sterilized PBS to a final concentration of 0.5 mg/ml.
Target Species Mouse
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.