Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse Tryptase beta-1/MCP-7/Mcpt7 Antibody, R&D Systems™
GREENER_CHOICE

Goat Polyclonal Antibody has been used in 4 publications

$200.50 - $561.50

Specifications

Antigen Tryptase beta-1/MCP-7/Mcpt7
Concentration LYOPH
Dilution Western Blot 0.1 ug/mL, Immunoprecipitation 25 ug/mL
Applications Western Blot, Immunoprecipitation
Classification Polyclonal
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
AF1937SP
GREENER_CHOICE
View Documents
R&D Systems
AF1937SP
25 μg
Each for $200.50
Only null left
Add to Cart
 
AF1937
GREENER_CHOICE
View Documents
R&D Systems
AF1937
100 μg
Each for $561.50
Only null left
Add to Cart
 
Description

Description

Tryptase beta-1/MCP-7/Mcpt7 Polyclonal specifically detects Tryptase beta-1/MCP-7/Mcpt7 in Mouse samples. It is validated for Western Blot, Immunoprecipitation.
Specifications

Specifications

Tryptase beta-1/MCP-7/Mcpt7
Western Blot 0.1 ug/mL, Immunoprecipitation 25 ug/mL
Polyclonal
Goat
Mouse
17230
Mouse myeloma cell line NS0-derived recombinant mouse Tryptase beta-1/MCP-7/Mcpt7 Ile29-Phe273 Accession # Q02844
Primary
Reconstitute at 0.2 mg/mL in sterile PBS.
Detects mouse Tryptase beta-1/MCP-7/Mcpt7 in direct ELISAs and Western blots. In direct ELISAs, approximately 10% cross-reactivity with recombinant human (rh) TPSB2 is observed, 5% cross-reactivity with rhTPS1 is observed, and less than 1% cross-reactivity with rhTPSG1 and recombinant mouse MCPT1 is observed.
LYOPH
Western Blot, Immunoprecipitation
Unconjugated
RUO
Q02844
Tpsab1
IgG
Affinity Purified
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.