Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Tryptase beta-2/TPSB2 Antibody (03), PerCP, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP306181PCP
Description
Tryptase beta-2/TPSB2 Monoclonal antibody specifically detects Tryptase beta-2/TPSB2 in Human samples. It is validated for ELISASpecifications
Tryptase beta-2/TPSB2 | |
Monoclonal | |
PerCP | |
PBS | |
Mouse | |
Protein A purified | |
RUO | |
Primary | |
Human | |
Purified |
ELISA | |
03 | |
ELISA | |
EC 3.4.21, EC 3.4.21.59, mast cell tryptase beta II, mast cell tryptase beta III, TPS2tryptase beta-2, tryptase beta 2, tryptase beta 2 (gene/pseudogene), tryptase beta II, tryptase beta III, Tryptase II, tryptase III, tryptase-2, tryptaseB, tryptaseC | |
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human Tryptase beta-2/TPSB2 (Accession#: NP_077078.5; Met 1-Pro 275). | |
0.1 mL | |
Cell Biology, Immunology, Innate Immunity | |
64499 | |
Store at 4C in the dark. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction