Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Tryptase gamma-1/TPSG1 Antibody (OTI1G1), mFluor Violet 450 SE, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP274652MFV450
Description
Tryptase gamma-1/TPSG1 Monoclonal antibody specifically detects Tryptase gamma-1/TPSG1 in Human samples. It is validated for Flow CytometrySpecifications
Tryptase gamma-1/TPSG1 | |
Monoclonal | |
mFluor Violet 450 SE | |
50mM Sodium Borate | |
Mouse | |
Immunogen affinity purified | |
RUO | |
25823 | |
Store at 4C in the dark. | |
IgG1 |
Flow Cytometry | |
OTI1G1 | |
Flow Cytometry | |
EC 3.4.21, EC 3.4.21.-, gamma I, gamma II, lung tryptase, mast cell protease II, mast cell tryptase, PRSS31skin tryptase, Serine protease 31, TMTpituitary tryptase, Transmembrane tryptase, trpA, tryptase gamma, tryptase gamma 1, tryptase gamma I, tryptase gamma II | |
Human recombinant protein fragment corresponding to amino acids 20-283 of human TPSG1(NP_036599) produced in E.coli. | |
0.1 mL | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction