Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Tryptase gamma-1/TPSG1 Antibody (OTI1E11), DyLight 650, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP274649C
Description
Tryptase gamma-1/TPSG1 Monoclonal specifically detects Tryptase gamma-1/TPSG1 in Human samples. It is validated for Western Blot, Flow Cytometry.Specifications
Tryptase gamma-1/TPSG1 | |
Monoclonal | |
DyLight 650 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
TPSG1 | |
Recombinant protein fragment corresponding to amino acids 20-283 of human Tryptase gamma-1/TPSG1 (NP_036599) produced in E.coli. | |
0.1 mL | |
25823.0 | |
Store at 4°C in the dark. |
Western Blot, Flow Cytometry | |
OTI1E11 | |
Western Blot, Flow Cytometry | |
EC 3.4.21, EC 3.4.21.-, gamma I, gamma II, lung tryptase, mast cell protease II, mast cell tryptase, PRSS31skin tryptase, Serine protease 31, TMTpituitary tryptase, Transmembrane tryptase, trpA, tryptase gamma, tryptase gamma 1, tryptase gamma I, tryptase gamma II | |
Mouse | |
Affinity Purified | |
Primary | |
Human | |
IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction