Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Tryptase Monoclonal Antibody (C10)
Mouse Monoclonal Antibody
$213.50 - $522.50
Specifications
Antigen | Tryptase |
---|---|
Clone | C10 |
Concentration | 1 mg/mL |
Content And Storage | -20°C |
Applications | Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
PIMA541687
|
Invitrogen™
MA541687 |
200 μL |
Each for $522.50
|
|
|||||
PIMA543557
|
Invitrogen™
MA543557 |
20 μL |
Each for $213.50
|
|
|||||
Description
Tryptase Monoclonal Antibody for Western Blot, ICC/IF, IHC (P)
Tryptase is a protein that belongs to the family of serine proteases and is primarily found in mast cells. It plays a crucial role in the immune response by cleaving and activating other proteins involved in inflammation and tissue repair. Tryptase is also involved in the regulation of blood pressure and has been implicated in various diseases, including asthma, allergies, and cardiovascular disorders. Its structure consists of four identical subunits that form a tetramer, and it is activated by proteolytic cleavage of its proenzyme form.Specifications
Tryptase | |
1 mg/mL | |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
Unconjugated | |
Mouse | |
RUO | |
PBS with 50% glycerol and 0.05% ProClin 300 | |
Q15661 | |
7177 | |
Recombinant protein Tryptase. The antigen corresponds to amino acid range 161-262 of the target protein. | |
Antibody |
C10 | |
-20°C | |
Monoclonal | |
Liquid | |
IgG2a κ | |
Human | |
Tpsab1 | |
TPSG1/TPSB2/TPSAB1/TPSD1 | |
Tpsab1 | |
Primary | |
Protein A/G |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title