Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Tryptic Soy Broth

Catalog No. R064882
Click to view available options
Quantity:
100/Pk.
20/Pk.
Format:
15mm x 103mm Tube
16mm x 125mm Tube
16mm x 125mm Tube, Hungate Cap
20mm x 113mm Tube
20mm x 150mm Tube
D - 1mL
D - 2mL
Tube
Fill Volume:
1 mL
10 mL
10 mL HC
15 mL
2 mL
20 mL
4.5 mL
5 mL
9 mL
This item is not returnable. View return policy
This item is not returnable. View return policy

Cultivate and isolate microorganisms with ease using this general purpose enrichment broth with formats available that meet USP guidelines for sterility testing.

Cultivate and isolate microorganisms with ease using Thermo Scientific™ Tryptic Soy Broth, a general purpose enrichment broth.

Remove the need for pipetting, dispensing or autoclaving with convenient, pre-filled and ready-to-use TSB tubes.

Part numbers beginning with R11 are for sterility testing according to USP (United States Pharmacopeia).

Not all products are available for sale in all territories. Please inquire.

Remel™ and Oxoid™ products are now part of the Thermo Scientific brand, combining powerful manual, semi-automated and fully automated test products and a comprehensive line of media and diagnostic products to offer a complete, end-to-end solution to quickly deliver the products you need and the quality results your laboratory depends on.
TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance AOAC, BAM, COMPF, SMWW, IDF, SMD, USDA, USP
Color Clear liquid, light to medium amber
Content And Storage 2 to 25°C
Description Tryptic Soy Broth
Format Tube
Manufacturing Origin United States
Product Type Microbiology Media
Fill Volume 1 mL
Packaging Type Glass tube with black screw cap
Quantity 20/Pk.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.