Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Tryptophan Hydroxylase 1/TPH-1 Antibody (007), mFluor Violet 500 SE, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP290021MFV500
Description
Tryptophan Hydroxylase 1/TPH-1 Monoclonal antibody specifically detects Tryptophan Hydroxylase 1/TPH-1 in Human samples. It is validated for ELISASpecifications
Tryptophan Hydroxylase 1/TPH-1 | |
Monoclonal | |
mFluor Violet 500 SE | |
50mM Sodium Borate | |
Rabbit | |
Protein A purified | |
RUO | |
Primary | |
Human | |
Purified |
ELISA | |
007 | |
ELISA | |
EC 1.14.16, EC 1.14.16.4, indoleacetic acid-5-hydroxylase, L-tryptophan hydroxylase, MGC119994, TPH, TPRH, TRPH, tryptophan 5-hydroxylase 1, Tryptophan 5-monooxygenase 1, tryptophan hydroxylase (tryptophan 5-monooxygenase), tryptophan hydroxylase 1 | |
Rabbit immunized with purified, recombinant Human Tryptophan Hydroxylase 1/TPH-1 (Uniprot#: P17752-1, Ile2-Ile444) | |
0.1 mL | |
Lipid and Metabolism, Neuroscience, Neurotransmission, Phospho Specific | |
7166 | |
Store at 4C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction