Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Tryptophan hydroxylase 2 Antibody, Alexa Fluor™ 594, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP177556AF594
Description
Tryptophan hydroxylase 2 Polyclonal specifically detects Tryptophan hydroxylase 2 in Human samples. It is validated for Western Blot.Specifications
| Tryptophan hydroxylase 2 | |
| Polyclonal | |
| Western Blot | |
| ADHD7, EC 1.14.16, EC 1.14.16.4, FLJ37295, MGC138872, Neuronal tryptophan hydroxylase, NTPHMGC138871, tryptophan 5-hydroxylase 2, Tryptophan 5-monooxygenase 2, tryptophan hydroxylase 2 | |
| Rabbit | |
| Affinity Purified | |
| Lipid and Metabolism, Phospho Specific | |
| 121278.0 | |
| Store at 4°C in the dark. |
| Western Blot | |
| Alexa Fluor 594 | |
| 50 mM sodium borate with 0.05% sodium azide | |
| TPH2 | |
| A synthetic peptide made to an N-terminal portion of the human Tryptophan hydroxylase 2 protein (between residues 1-100) [Uniprot: Q8IWU9] | |
| 0.1 mL | |
| Primary | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction