Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tryptophan Hydroxylase Pan Specific Antibody, R&D Systems™

Catalog No. PPS038
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PPS038 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PPS038 Supplier R&D Systems Supplier No. PPS038
This item is not returnable. View return policy

Sheep Polyclonal Antibody

Prepared from sheep serum by affinity purification using a column to which the recombinant protein immunogen was coupled.

Tryptophan Hydroxylase Polyclonal specifically detects Tryptophan Hydroxylase in Multi-Species samples. It is validated for Western Blot, Immunohistochemistry.

Specifications

Antigen Tryptophan Hydroxylase
Applications Western Blot, Immunohistochemistry
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:1000 dilution, Immunohistochemistry 1:1000 dilution
Host Species Sheep
Immunogen Tryptophan Hydroxylase
Purification Method Affinity Purified
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity This antibody is specific for the ∼55 kDa TPH in Western blots of human dorsal Raphe nucleus lysates. Immunolabeling has been demonstrated in rat brain lysates. Based upon the relatively high degree of homology of tryptophan hydroxylase, the antibodies should cross-react with other mammalian species. Due to antigen modifications in E. coli, this antibody does not recognize TPH in rabbit tissues.
Target Species Multi-species
Content And Storage For long-term storage, < -20degrees C is recommended. Product is stable at < -20degrees C for at least 1 year.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.