Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TPH1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PA14678
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PA14678 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PA14678 Supplier Invitrogen™ Supplier No. PA14678
Add to Cart
Add to Cart

Sheep Polyclonal Antibody

This antibody is predicted to react with wide variety of mammals based on 100% sequence homology, however it does not recognize TPH in rabbit tissues. This antibody is specific for the ~55 kDa TPH protein in Western blots of human dorsal Raphe nucleus. Immunolabeling has been demonstrated in rat brain extracts.

Tryptophan hydroxylase (TPH) catalyzes the 5-hydroxylation of tryptophan, which is the first step in the biosynthesis of indoleamines (serotonin and melatonin) (Martinez et al., 2001). In mammals, serotonin biosynthesis occurs predominantly in neurons which originate in the Raphe nuclei of the brain, and melatonin synthesis takes place within the pineal gland. Although TPH catalyzes the same reaction within the Raphe nuclei and the pineal gland, TPH activity is rate-limiting for serotonin but not melatonin biosynthesis. Serotonin functions mainly as a neurotransmitter, whereas melatonin is the principal hormone secreted by the pineal gland. The activity of TPH is enhanced by phosphorylation by cAMP-dependent protein kinase (PKA) and Ca2+/calmodulin kinase II (CAM K II) (Jiang et al., 2000; Johansen et al., 1996). Both PKA and CAM K II phosphorylate Ser58 which lies within the regulatory domain of TPH (Kuhn et al., 1997).
TRUSTED_SUSTAINABILITY

Specifications

Antigen TPH1
Applications Immunohistochemistry, Immunohistochemistry (Frozen), Western Blot
Classification Polyclonal
Concentration 0.17 mg/mL
Conjugate Unconjugated
Formulation 0.01M HEPES with 100μg/mL BSA, 50% glycerol, 0.15M NaCl and no preservative; pH 7.5
Gene TPH1
Gene Accession No. P09810, P17290, P17752
Gene Alias indoleacetic acid-5-hydroxylase; L-tryptophan hydroxylase; MGC119994; TPH; Tph1; TPH2; TPRH; TRPH; tryptophan 5-hydroxylase 1; Tryptophan 5-monooxygenase 1; tryptophan hydroxylase; tryptophan hydroxylase (tryptophan 5-monooxygenase); tryptophan hydroxylase 1; tryptophan hydroxylase 1 (tryptophan 5-monooxygenase)
Gene Symbols TPH1
Host Species Sheep
Immunogen Recombinant rabbit tryptophan hydroxylase, isolated as inclusion bodies from E. coli and purified by preparative SDS-PAGE.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100009100, 100101558, 24848, 7166
Target Species Human, Rat, Rabbit
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.