Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TPH1 Recombinant Rabbit Monoclonal Antibody (145)
GREENER_CHOICE

Catalog No. PIMA529618
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA529618 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA529618 Supplier Thermo Scientific Supplier No. MA529618
Add to Cart
Add to Cart

TPH1 Recombinant Rabbit Monoclonal Antibody (145)

TPH1 Recombinant Monoclonal Antibody for Western Blot, IP

TRUSTED_SUSTAINABILITY

Specifications

Antigen TPH1
Applications Immunoprecipitation, Western Blot
Classification Recombinant Monoclonal
Clone 145
Concentration 5.0 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative
Gene TPH1
Gene Accession No. P17752
Gene Alias indoleacetic acid-5-hydroxylase; L-tryptophan hydroxylase; MGC119994; TPH; Tph1; TPH2; TPRH; TRPH; tryptophan 5-hydroxylase 1; Tryptophan 5-monooxygenase 1; tryptophan hydroxylase; tryptophan hydroxylase (tryptophan 5-monooxygenase); tryptophan hydroxylase 1; tryptophan hydroxylase 1 (tryptophan 5-monooxygenase)
Gene Symbols TPH1
Host Species Rabbit
Immunogen Recombinant Human Tryptophan Hydroxylase 1/TPH1 Protein (Ile2-Ile444).
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7166
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.