Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tryptophan rich protein Antibody, Alexa Fluor™ 488, Novus Biologicals™
SDP

Rabbit Polyclonal Antibody

Supplier:  Novus Biologicals NBP297813AF488

Catalog No. NB048042


Only null left
Add to Cart

Description

Description

Tryptophan rich protein Polyclonal specifically detects Tryptophan rich protein in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications

Specifications

Tryptophan rich protein
Polyclonal
Immunohistochemistry, Immunohistochemistry-Paraffin
congenital heart disease 5 protein10tryptophan-rich protein, FLJ51808, tryptophan rich basic protein
Produced in rabbits immunized with E. coli-derived Human Tryptophan rich protein fragment.
0.1 mL
Primary
Human
Purified
Immunohistochemistry, Immunohistochemistry (Paraffin)
Alexa Fluor 488
50mM Sodium Borate
Rabbit
Antigen Affinity-purified
RUO
7485
Store at 4°C in the dark.
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.