Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Tryptophan rich protein Antibody, PerCP, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP297813PCP
Description
Tryptophan rich protein Polyclonal antibody specifically detects Tryptophan rich protein in Human samples. It is validated for Immunohistochemistry (Paraffin)Specifications
Tryptophan rich protein | |
Polyclonal | |
Immunohistochemistry-Paraffin | |
congenital heart disease 5 protein10tryptophan-rich protein, FLJ51808, tryptophan rich basic protein | |
Produced in rabbits immunized with E. coli-derived Human Tryptophan rich protein fragment. | |
0.1 mL | |
Neurodegeneration, Neuroscience | |
7485 | |
Store at 4C in the dark. | |
IgG |
Immunohistochemistry (Paraffin) | |
PerCP | |
PBS | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction