Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Tryptophanyl tRNA synthetase Antibody, PerCP, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP232185PCP
Description
Tryptophanyl tRNA synthetase Polyclonal antibody specifically detects Tryptophanyl tRNA synthetase in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, ImmunoprecipitationSpecifications
Tryptophanyl tRNA synthetase | |
Polyclonal | |
Western Blot, Immunohistochemistry, Immunoprecipitation | |
EC 6.1.1, EC 6.1.1.2, GAMMA-2, hWRS, IFI53, IFP53trpRS, Interferon-induced protein 53, TrpRS, tryptophan tRNA ligase 1, cytoplasmic, Tryptophan--tRNA ligase, tryptophanyl-tRNA synthetase, tryptophanyl-tRNA synthetase, cytoplasmic, WRS | |
The immunogen this antibody was made to, maps to a region between residue 1 to 50 of human Tryptophanyl-tRNA Synthetase using the numbering given in entry NP_004175.2 | |
0.1 mL | |
Cell Cycle and Replication | |
7453 | |
Store at 4C in the dark. | |
IgG |
Western Blot, Immunohistochemistry, Immunoprecipitation | |
PerCP | |
PBS | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human, Mouse | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction