Learn More
Description
The basis for signal amplification comes from the specific and high-density deposition of ClariTSA dyes adjacent to a horseradish peroxidase (HRP)-labeled probe in a process termed Tyramide Signal Amplification (TSA) or Catalyzed Reporter Deposition (CARD).
Excitation maximum = 654 nm; emission maximum = 668 nm
Please see the product datasheet or protocol for information on product use.
Specifications
Specifications
| Quantity | 1 Kit (20 to 80 Slides) |
| Product Line | Tocris |
| Target | Tyramide Signaling Amplification/TSA |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

