Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TSARG2 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501981255
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-1255 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-1255 Supplier Bioss Supplier No. BS11309R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The testis spermatocyte apoptosis-related gene 2 protein (TSARG2, also designated spermatogenesis associated-4 or SPATA4) is involved in spermatogenesis. TSARG2 is specifically expressed in spermatogonia and spermatocytes of the seminiferous tubules, and it localizes to the nucleus. The predicted molecular weight of TSARG2 ranges depending on the species. TSARG2 is significantly upregulated in cryptorchidism and therefore, is a testis-specific apoptosis candidate oncogene.

Specifications

Antigen TSARG2
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene SPATA4
Gene Accession No. Q8NEY3
Gene Alias 1700001N01Rik; SPATA4; SPEF1B; spermatogenesis associated 4; spermatogenesis related gene 2; spermatogenesis-associated 4; Spermatogenesis-associated protein 4; Spermatogenesis-related gene 2 protein; Srg2; testis and spermatogenesis cell related protein 2; Testis and spermatogenesis cell-related protein 2; Testis spermatocyte apoptosis-related gene 2 protein; TSARG2
Gene Symbols SPATA4
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human TSARG2.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 132851
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.