Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TSARG7 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501984297
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-4297 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-4297 Supplier Bioss Supplier No. BS1924R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Lysophosphatidic acid acyltransferases (EC 2.3.1.51) catalyze the conversion of lysophosphatidic acid (LPA) to phosphatidic acid (PA). LPA and PA are involved in signal transduction and lipid biosynthesis.

Specifications

Antigen TSARG7
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene GPAT4
Gene Accession No. Q86UL3
Gene Alias 1-acylglycerol-3-phosphate O-acyltransferase 6; 1-acylglycerol-3-phosphate O-acyltransferase 6 (lysophosphatidic acid acyltransferase, zeta); 1-acyl-sn-glycerol-3-phosphate acyltransferase zeta; 1-AGP; 1-AGP acyltransferase 6; 1-AGPAT 6; Acyl-CoA:glycerol-3-phosphate acyltransferase 4; Agpat6; AU041707; AW545732; Glycerol-3-phosphate acyltransferase 4; glycerol-3-phosphate acyltransferase 6; glycerol-3-phosphate acyltransferase-4; Gpat4; LPAATZ; LPAAT-zeta; Lysophosphatidic acid acyltransferase zeta; putative lysophosphatidic acid acyltransferase; RGD1310520; Testis spermatogenesis apoptosis-related protein 7; Tsarg7; UNQ551/PRO1108
Gene Symbols GPAT4
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human TSARG7.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 137964
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.