Learn More
TSC1 (Hamartin) Monoclonal Antibody (5C8A12), Abbomax
Mouse Monoclonal Antibody
Supplier: Abbomax Inc 606380

Description
Positive control: Breast carcinoma Cellular location: Secreted.
TSC1 (Hamartin) Monoclonal antibody specifically detects TSC1 (Hamartin) in Human, Mouse, Rat samples. It is validated for Immunohistochemistry (Paraffin)Specifications
TSC1 (Hamartin) | |
Monoclonal | |
0.5 mg/mL | |
PBS with proprietary stabilizer and 0.01% sodium azide | |
Q92574, Q9EP53, Q9Z136 | |
Mouse | |
Antigen affinity chromatography | |
RUO | |
60445, 64930, 7248 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
5C8A12 | |
Unconjugated | |
TSC1 | |
hamartin; HGNC:12362; Kiaa0243; LAM; MGC86987; OTTHUMP00000022439; OTTHUMP00000198378; TSC; Tsc1; tuberous sclerosis 1; tuberous sclerosis 1 protein; Tuberous sclerosis 1 protein homolog; tumor suppressor | |
A synthetic peptide conrresponding to internal segment of human TSC1 | |
100 μg | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.