Learn More
Invitrogen™ TSC1 Recombinant Rabbit Monoclonal Antibody (6Q10T10)

Description
Immunogen sequence: TTMGEASASI PTTVGSLPSS KSFLGMKARE LFRNKSESQC DEDGMTSSLS ESLKTELGKD LGVEAKIPLN LDGPHPSPPT PDSVGQLHIM DYNETHHEHS.
Specifications
Specifications
| Antigen | TSC1 |
| Applications | ELISA, Western Blot |
| Classification | Recombinant Monoclonal |
| Clone | 6Q10T10 |
| Concentration | 0.49 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.05% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 |
| Gene | TSC1 |
| Gene Accession No. | Q92574 |
| Gene Alias | hamartin; HGNC:12362; Kiaa0243; LAM; MGC86987; OTTHUMP00000022439; OTTHUMP00000198378; TSC; Tsc1; tuberous sclerosis 1; tuberous sclerosis 1 protein; Tuberous sclerosis 1 protein homolog; tumor suppressor |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.