Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TSC1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA520131
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA520131 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA520131 Supplier Invitrogen™ Supplier No. PA520131
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This antibody recognizes both isoforms of TSC1. A suggested positive control is EL4 cell lysate. PA5-20131 can be used with blocking peptide PEP-0249.

Tuberous sclerosis complex (TSC) is an autosomal dominant tumor syndrome caused by mutations in either of the TSC1 or TSC2 tumor suppressor genes. The products of these genes form a protein complex that indirectly decreases the signaling of the mammalian Target of Rapamycin (TOR), an evolutionarily conserved serine/threonine kinase that regulates cell growth and cell cycle through its ability to integrate signals from nutrient levels and growth factors. TOR activity is stimulated by Rheb, a member of the Ras superfamily of G-proteins, when the GTP/GDP ratio bound to Rheb is high. Immunoprecipitated TSC1/TSC2 has been shown to stimulate Rheb GTPase activity in vitro, suggesting that the TSC1/TSC2 decreases the ability of Rheb to stimulate TOR activity. This is supported by experiments showing overexpression of TSC1 and TSC2 results in a significant decrease in the GTP/GDP ratio bound to Rheb and the inhibition of cell growth. A shorter 40 kDa isoform of TSC1 has been shown to exist but its function is unknown.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TSC1
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.02% sodium azide
Gene TSC1
Gene Accession No. Q92574, Q9EP53, Q9Z136
Gene Alias hamartin; HGNC:12362; Kiaa0243; LAM; MGC86987; OTTHUMP00000022439; OTTHUMP00000198378; TSC; Tsc1; tuberous sclerosis 1; tuberous sclerosis 1 protein; Tuberous sclerosis 1 protein homolog; tumor suppressor
Gene Symbols TSC1
Host Species Rabbit
Immunogen A 15 amino acid synthetic peptide from the middle region of human TSC1.
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 60445, 64930, 7248
Target Species Human, Mouse, Rat
Content And Storage Maintain refrigerated at 2-8°C for up to 3 months. For long term storage store at -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.