Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TSC22 domain family, member 4 (TSC22D4) Monoclonal Antibody (OTI3G6), TrueMAB™, OriGene
SDP

Catalog No. 502543937
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-254-3937 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-254-3937 Supplier OriGene Supplier No. CF811733
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

TSC22 domain family, member 4 (TSC22D4) Monoclonal antibody specifically detects TSC22 domain family, member 4 (TSC22D4) in Human samples. It is validated for Western Blot

Specifications

Antigen TSC22 domain family, member 4 (TSC22D4)
Applications Western Blot
Classification Monoclonal
Clone OTI3G6
Concentration See Label
Conjugate Unconjugated
Formulation PBS with 8% trehalose and no preservative; pH 7.3
Gene FANCC
Gene Accession No. Q00597
Gene Alias 0610009M14Rik; 1700023B23Rik; AI415410; LOW QUALITY PROTEIN: TSC22 domain family protein 4; THG1; THG-1; Thg1pit; Thg-1pit; Tilz2; TSC22 domain family 4; TSC22 domain family member 4; TSC22 domain family protein 4; TSC22 domain family, member 4; Tsc22d4; tsc-22-like protein THG-1; TSC22-related inducible leucine zipper 2; TSC22-related-inducible leucine zipper protein 2
Host Species Mouse
Immunogen Human recombinant protein fragment corresponding to amino acids 1-284 of human FANCC produced in E.coli.
Purification Method Protein A/G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2176
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Form Lyophilized
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.