Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TSC22/TSC22D1 Antibody (110), mFluor Violet 450 SE, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP290144MFV450
Description
TSC22/TSC22D1 Monoclonal antibody specifically detects TSC22/TSC22D1 in Human samples. It is validated for Immunocytochemistry/ ImmunofluorescenceSpecifications
TSC22/TSC22D1 | |
Monoclonal | |
mFluor Violet 450 SE | |
50mM Sodium Borate | |
Rabbit | |
Protein A purified | |
RUO | |
Primary | |
Human | |
Purified |
Immunocytochemistry | |
110 | |
Immunocytochemistry/ Immunofluorescence | |
Cerebral protein 2, KIAA1994, MGC17597, Regulatory protein TSC-22, RP11-269C23.2, TGFB1I4, TGFB-stimulated clone 22 homolog, transforming growth factor beta 1 induced transcript 4, Transforming growth factor beta-1-induced transcript 4 protein, transforming growth factor beta-stimulated protein TSC-22, TSC22 domain family 1, TSC22 domain family protein 1, TSC22 domain family, member 1, TSC22DKFZp686O19206 | |
This antibody was obtained from a rabbit immunized with purified, recombinant Human TSC22/TSC22D1 (Uniprot#: Q15714-2; Met1-Ala144). | |
0.1 mL | |
Cancer | |
8848 | |
Store at 4C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction