Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TSC22/TSC22D1 Antibody, Novus Biologicals™
SDP

Rabbit Polyclonal Antibody

$181.90 - $402.00

Specifications

Antigen TSC22/TSC22D1
Concentration 0.5 mg/mL
Dilution Western Blot 3-5 ug/ml
Applications Western Blot
Classification Polyclonal
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NBP224628SS
SDP
View Documents
Novus Biologicals
NBP224628SS
0.025 mg
Each for $181.90
Only null left
Add to Cart
 
NBP224628
SDP
View Documents
Novus Biologicals
NBP224628
0.1 mg
Each for $402.00
Only null left
Add to Cart
 
Description

Description

TSC22/TSC22D1 Polyclonal specifically detects TSC22/TSC22D1 in Human, Mouse, Rat, Canine samples. It is validated for Western Blot.
Specifications

Specifications

TSC22/TSC22D1
Western Blot 3-5 ug/ml
Polyclonal
Purified
RUO
Human, Mouse, Rat, Canine
Cerebral protein 2, KIAA1994, MGC17597, Regulatory protein TSC-22, RP11-269C23.2, TGFB1I4, TGFB-stimulated clone 22 homolog, transforming growth factor beta 1 induced transcript 4, Transforming growth factor beta-1-induced transcript 4 protein, transforming growth factor beta-stimulated protein TSC-22, TSC22 domain family 1, TSC22 domain family protein 1, TSC22 domain family, member 1, TSC22DKFZp686O19206
TSC22D1
IgG
Protein A purified
This antibody may also recognize TSC22D2 as the TSC22D1 sequence used for immunogen is 87% homologous to TSC22D2.
0.5 mg/mL
Western Blot
Unconjugated
Rabbit
Cancer
0.2 ml PBS and 0.05% BSA with 0.05% Sodium Azide
8848
A portion of amino acids 100-150 of human TSC22 was used as the immunogen.
Primary
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.