Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TSC22/TSC22D1 Rabbit anti-Human, Clone: 143, Novus Biologicals™

Rabbit Monoclonal Antibody
$353.50 - $560.50
Specifications
Antigen | TSC22/TSC22D1 |
---|---|
Clone | 143 |
Dilution | Immunocytochemistry/Immunofluorescence 1:20-1:100, Immunofluorescence |
Applications | Immunofluorescence |
Classification | Monoclonal |
Description
TSC22/TSC22D1 Monoclonal specifically detects TSC22/TSC22D1 in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.Specifications
TSC22/TSC22D1 | |
Immunocytochemistry/Immunofluorescence 1:20-1:100, Immunofluorescence | |
Monoclonal | |
Purified | |
RUO | |
Human | |
Cerebral protein 2, KIAA1994, MGC17597, Regulatory protein TSC-22, RP11-269C23.2, TGFB1I4, TGFB-stimulated clone 22 homolog, transforming growth factor beta 1 induced transcript 4, Transforming growth factor beta-1-induced transcript 4 protein, transforming growth factor beta-stimulated protein TSC-22, TSC22 domain family 1, TSC22 domain family protein 1, TSC22 domain family, member 1, TSC22DKFZp686O19206 | |
Recombinant Human TSC22/TSC22D1 protein | |
Primary | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
143 | |
Immunofluorescence | |
Unconjugated | |
Rabbit | |
Cancer | |
0.2 um filtered solution in PBS | |
8848 | |
IgG | |
Protein A purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title