Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TSC22D1 Monoclonal Antibody (OTI4A11), TrueMAB™, OriGene

Mouse Monoclonal Antibody
Supplier: OriGene TA505061S


Description
TSC22D1 Monoclonal antibody specifically detects TSC22D1 in Human samples. It is validated for Immunohistochemistry (Paraffin), Western BlotSpecifications
TSC22D1 | |
Monoclonal | |
1 mg/mL | |
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
Q15714 | |
Mouse | |
Affinity chromatography | |
RUO | |
8848 | |
-20° C, Avoid Freeze/Thaw Cycles | |
IgG1 |
Immunohistochemistry (Paraffin), Western Blot | |
OTI4A11 | |
Unconjugated | |
Tsc22d1 | |
AA589566; AW105905; Cerebral protein 2; Egr5; hucep-2; Kiaa1994; Ptg-2; regulatory protein Tsc22; Regulatory protein TSC-22; RP11-269C23.2; short tandem repeat locus PEZ20 variant 19; Tgfb1i4; TGFbeta-stimulated clone 22; Tgfb-stimulated clone 22 homolog; Tilz1b; transcriptional regulator TSC-22; transforming growth factor beta 1 induced transcript 4; Transforming growth factor beta stimulated clone 22; transforming growth factor beta-1-induced transcript 4 protein; transforming growth factor beta-stimulated protein TSC-22; TS22A; Tsc; Tsc22; TSC-22; TSC22 domain family member 1; TSC22 domain family protein 1; TSC22 domain family, member 1; TSC22D1; Tsc22-related inducible leucine zipper 1b | |
Full length human recombinant protein of human TSC22D1 produced in HEK293T cell. | |
30 μL | |
Primary | |
Human | |
Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction