Learn More
Invitrogen™ TSC22D1 Monoclonal Antibody (OTI4H8)

Description
TSC22D1 Monoclonal Antibody for Western Blot, IHC (P), Flow
Specifications
Specifications
| Antigen | TSC22D1 |
| Applications | Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot |
| Classification | Monoclonal |
| Clone | OTI4H8 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 |
| Gene | Tsc22d1 |
| Gene Accession No. | Q15714 |
| Gene Alias | AA589566; AW105905; Cerebral protein 2; Egr5; hucep-2; Kiaa1994; Ptg-2; regulatory protein Tsc22; Regulatory protein TSC-22; RP11-269C23.2; short tandem repeat locus PEZ20 variant 19; Tgfb1i4; TGFbeta-stimulated clone 22; Tgfb-stimulated clone 22 homolog; Tilz1b; transcriptional regulator TSC-22; transforming growth factor beta 1 induced transcript 4; Transforming growth factor beta stimulated clone 22; transforming growth factor beta-1-induced transcript 4 protein; transforming growth factor beta-stimulated protein TSC-22; TS22A; Tsc; Tsc22; TSC-22; TSC22 domain family member 1; TSC22 domain family protein 1; TSC22 domain family, member 1; TSC22D1; Tsc22-related inducible leucine zipper 1b |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.