Learn More
Invitrogen™ TSC22D1 Polyclonal Antibody

Description
The amino acid sequence used as immunogen is 86% homologous in Zebrafish.
Specifications
Specifications
| Antigen | TSC22D1 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.05% BSA and 0.05% sodium azide |
| Gene | Tsc22d1 |
| Gene Accession No. | P62500, P62501, Q15714 |
| Gene Alias | AA589566; AW105905; Cerebral protein 2; Egr5; hucep-2; Kiaa1994; Ptg-2; regulatory protein Tsc22; Regulatory protein TSC-22; RP11-269C23.2; short tandem repeat locus PEZ20 variant 19; Tgfb1i4; TGFbeta-stimulated clone 22; Tgfb-stimulated clone 22 homolog; Tilz1b; transcriptional regulator TSC-22; transforming growth factor beta 1 induced transcript 4; Transforming growth factor beta stimulated clone 22; transforming growth factor beta-1-induced transcript 4 protein; transforming growth factor beta-stimulated protein TSC-22; TS22A; Tsc; Tsc22; TSC-22; TSC22 domain family member 1; TSC22 domain family protein 1; TSC22 domain family, member 1; TSC22D1; Tsc22-related inducible leucine zipper 1b |
| Gene Symbols | Tsc22d1 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.