Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TSC22D3 Monoclonal Antibody (OTI1F11), TrueMAB™, OriGene
SDP

Catalog No. 502539799
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
30 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-253-9799 30 μL
50-167-5797 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-253-9799 Supplier OriGene Supplier No. TA505293S
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

TSC22D3 Monoclonal antibody specifically detects TSC22D3 in Human samples. It is validated for Western Blot

Specifications

Antigen TSC22D3
Applications Western Blot
Classification Monoclonal
Clone OTI1F11
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene SRY
Gene Accession No. Q05066
Gene Alias delta sleep inducing peptide, immunoreactor; delta sleep-inducing peptide immunoreactor; DIP; Dsip1; DSIPI; DSIP-immunoreactive leucine zipper protein; DSIP-immunoreactive peptide; GILZ; glucocorticoid-induced leucine zipper protein; long glucocorticoid-induced leucine zipper protein; Protein DIP; Tilz3; TSC22 domain family 3; TSC22 domain family member 3; TSC22 domain family protein 3; TSC22 domain family, member 3; TSC-22 related protein; TSC22D3; TSC-22-like protein; TSC-22R; TSC22-related inducible leucine zipper 3; TSC-22-related protein; TSC22-related-inducible leucine zipper 3
Host Species Mouse
Immunogen Full length human recombinant protein of human SRY produced in E.coli.
Purification Method Affinity chromatography
Quantity 30 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6736
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.