Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ TSC22D4 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA540333
Description
Peptide sequence: SGGKKKSSFQ ITSVTTDYEG PGSPGASDPP TPQPPTGPPP RLPNGEPSPD Sequence homology: Dog: 90%; Guinea Pig: 93%; Human: 100%; Rat: 77%.
TSC22D4 is a member of the TSC22 domain family of leucine zipper transcriptional regulators. Gene Ontology (GO) annotations related to this gene include DNA-binding transcription factor activity. An important paralog of this gene is TSC22D1. Binds DNA and acts as a transcriptional repressor (PubMed:10488076). Involved in the regulation of systematic glucose homeostasis and insulin sensitivity, via transcriptional repression of downstream insulin signaling targets such as OBP2A/LCN13 (By similarity). Acts as a negative regulator of lipogenic gene expression in hepatocytes and thereby mediates the control of very low-density lipoprotein release (PubMed:23307490). May play a role in neurite elongation and survival (By similarity).
Specifications
| TSC22D4 | |
| Polyclonal | |
| Unconjugated | |
| Tsc22d4 | |
| 0610009M14Rik; 1700023B23Rik; AI415410; LOW QUALITY PROTEIN: TSC22 domain family protein 4; THG1; THG-1; Thg1pit; Thg-1pit; Tilz2; TSC22 domain family 4; TSC22 domain family member 4; TSC22 domain family protein 4; TSC22 domain family, member 4; Tsc22d4; tsc-22-like protein THG-1; TSC22-related inducible leucine zipper 2; TSC22-related-inducible leucine zipper protein 2 | |
| Rabbit | |
| Protein A | |
| RUO | |
| 81628 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 1 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q9Y3Q8 | |
| Tsc22d4 | |
| Synthetic peptide directed towards the N-terminal of human TSC22D4 (aa 2-51). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction