Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TSC22D4 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA568853
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA568853 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA568853 Supplier Invitrogen™ Supplier No. PA568853
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This target displays homology in the following species: Cow: 79%; Dog: 77%; Guinea Pig: 92%; Horse: 86%; Human: 92%; Mouse: 100%; Pig: 93%; Rabbit: 86%; Rat: 100%.

TSC22D4 is a member of the TSC22 domain family of leucine zipper transcriptional regulators. Gene Ontology (GO) annotations related to this gene include DNA-binding transcription factor activity. An important paralog of this gene is TSC22D1. Binds DNA and acts as a transcriptional repressor (PubMed:10488076). Involved in the regulation of systematic glucose homeostasis and insulin sensitivity, via transcriptional repression of downstream insulin signaling targets such as OBP2A/LCN13 (By similarity). Acts as a negative regulator of lipogenic gene expression in hepatocytes and thereby mediates the control of very low-density lipoprotein release (PubMed:23307490). May play a role in neurite elongation and survival (By similarity).
TRUSTED_SUSTAINABILITY

Specifications

Antigen TSC22D4
Applications Immunohistochemistry, Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene Tsc22d4
Gene Accession No. Q9EQN3
Gene Alias 0610009M14Rik; 1700023B23Rik; AI415410; LOW QUALITY PROTEIN: TSC22 domain family protein 4; THG1; THG-1; Thg1pit; Thg-1pit; Tilz2; TSC22 domain family 4; TSC22 domain family member 4; TSC22 domain family protein 4; TSC22 domain family, member 4; Tsc22d4; tsc-22-like protein THG-1; TSC22-related inducible leucine zipper 2; TSC22-related-inducible leucine zipper protein 2
Gene Symbols Tsc22d4
Host Species Rabbit
Immunogen synthetic peptide directed towards the N terminal region of mouse TSC22D4.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 78829
Target Species Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.