Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ TSC22D4 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA568854
Description
This target displays homology in the following species: Cow: 90%; Guinea Pig: 90%; Human: 90%; Mouse: 100%; Rabbit: 90%; Rat: 100%.
TSC22D4 is a member of the TSC22 domain family of leucine zipper transcriptional regulators. Gene Ontology (GO) annotations related to this gene include DNA-binding transcription factor activity. An important paralog of this gene is TSC22D1. Binds DNA and acts as a transcriptional repressor (PubMed:10488076). Involved in the regulation of systematic glucose homeostasis and insulin sensitivity, via transcriptional repression of downstream insulin signaling targets such as OBP2A/LCN13 (By similarity). Acts as a negative regulator of lipogenic gene expression in hepatocytes and thereby mediates the control of very low-density lipoprotein release (PubMed:23307490). May play a role in neurite elongation and survival (By similarity).
Specifications
TSC22D4 | |
Polyclonal | |
Unconjugated | |
Tsc22d4 | |
0610009M14Rik; 1700023B23Rik; AI415410; LOW QUALITY PROTEIN: TSC22 domain family protein 4; THG1; THG-1; Thg1pit; Thg-1pit; Tilz2; TSC22 domain family 4; TSC22 domain family member 4; TSC22 domain family protein 4; TSC22 domain family, member 4; Tsc22d4; tsc-22-like protein THG-1; TSC22-related inducible leucine zipper 2; TSC22-related-inducible leucine zipper protein 2 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
78829 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q9EQN3 | |
Tsc22d4 | |
Synthetic peptide directed towards the middle region of mouse TSC22D4 (aa 241-290). | |
100 μL | |
Primary | |
Mouse | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction