Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TSC22D4 Rabbit anti-Human, Polyclonal, Bethyl Laboratories

Catalog No. 501564577
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-156-4577 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-156-4577 Supplier Bethyl Laboratories, Inc Supplier No. A303222A
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The recommended shelf life for this product is 1 year from date of receipt.

TSC22D4 is a leucine zipper-containing protein that is highly conserved during evolution. It is transcriptionally up-regulated by many different stimuli, including anti-cancer drugs and growth inhibitors. TSC22D4 may play a suppressive role in tumorigenesis.

Specifications

Antigen TSC22D4
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/ml
Conjugate Unconjugated
Formulation phosphate, tris citrate with 0.09% sodium azide; pH 7-8
Gene TSC22D4
Gene Accession No. Q9Y3Q8
Gene Alias TSC22 domain family member 4, THG1, THG-1, TILZ2
Gene Symbols TSC22D4
Host Species Rabbit
Immunogen Between 345 and 395
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 81628
Target Species Human
Content And Storage 4° C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.