Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TSCOT Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA590509
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA590509 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA590509 Supplier Invitrogen™ Supplier No. PA590509
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MSPEVTCPRR GHLPRFHPRT WVEPVVASSQ VAASLYDAGL LLVVKASYGT GGSSNHSASP SPRGALEDQQ QRAISNFYII YNLVVGLSPL LSAYGLGWLS DRYHRKISIC; Positive Samples: LO2, Raji, MCF7, Mouse heart, Mouse brain, Mouse spleen; Cellular Location: Cell membrane, Multi-pass membrane protein.

The TSCOT gene, known as the Thymic Stromal Co-transporter, plays a crucial role in the gene expression regulation within thymic epithelial cells, essential for T cell development. It encodes a protein that bears weak homology to bacterial 12 transmembrane co-transporters. Research indicates that TSCOT is predominantly expressed in thymic tissues, although low levels of expression are detectable in other epithelial tissues such as skin and lungs. The gene has been co-analyzed with Foxn1 and Hoxa3, highlighting its significant role in organogenesis. Studies utilizing competitive reverse transcription-polymerase chain reaction (RT-PCR) have mapped TSCOT to specific genomic regions, verifying its expression in a tissue and cell-type specific manner. The introduction of a neoantigen into TSCOT-expressing cells demonstrates its capability to mediate antigen-specific T cell tolerance, offering potential applications in immunological therapies.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TSCOT
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 1.47 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene SLC46A2
Gene Accession No. Q8CA03, Q9BY10
Gene Alias 5430429N04Rik; Ly110; putative thymic stromal co-transporter TSCOT; SLC46A2; Solute carrier family 46 member 2; solute carrier family 46, member 2; thymic stromal co-transporter; thymic stromal cotransporter homolog; Thymic stromal cotransporter protein; TSCOT; TSO-1C12
Gene Symbols SLC46A2
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-110 of human SLC46A2 (NP_1490403).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 30936, 57864
Target Species Human, Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.