Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TSEN2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA596827
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA596827 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA596827 Supplier Invitrogen™ Supplier No. PA596827
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MAEAVFHAPK RKRRVYETYE SPLPIPFGQD HGPLKEFKIF RAEMINNNVI VRNAEDIEQL YGKGYFGKGI LSRSRPSFTI SDPKLVAKWK DMKTNMPIIT SKRYQHSVEW AAELMRRQGQ DESTVRRILK DYTKPLEHPP VKRNEEAQVH DKLNSGMVSN MEGTAGGERP SVVNGDSGKS GGVGDPREPL GCLQEGSGCH PTTESFEKSV REDASPLPHV; Positive Samples: MCF-7, HeLa, A-549, Mouse heart, Mouse brain, Rat heart; Cellular Location: Nucleus, nucleolus.

Constitutes one of the two catalytic subunit of the tRNA-splicing endonuclease complex, a complex responsible for identification and cleavage of the splice sites in pre-tRNA. It cleaves pre-tRNA at the 5'- and 3'-splice sites to release the intron. The products are an intron and two tRNA half-molecules bearing 2',3'-cyclic phosphate and 5'-OH termini. There are no conserved sequences at the splice sites, but the intron is invariably located at the same site in the gene, placing the splice sites an invariant distance from the constant structural features of the tRNA body. Isoform 1 probably carries the active site for 5'-splice site cleavage. The tRNA splicing endonuclease is also involved in mRNA processing via its association with pre- mRNA 3'-end processing factors, establishing a link between pre- tRNA splicing and pre-mRNA 3'-end formation, suggesting that the endonuclease subunits function in multiple RNA-processing events. Isoform 2 is responsible for processing a yet unknown RNA substrate. The complex containing isoform 2 is not able to cleave pre-tRNAs properly, although it retains endonucleolytic activity.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TSEN2
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 1.11 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene Tsen2
Gene Accession No. Q5M954, Q6P7W5, Q8NCE0
Gene Alias AU067695; HsSen2; PCH2B; RGD1309946; SEN2; SEN2L; tRNA splicing endonuclease 2 homolog (S. cerevisiae); tRNA splicing endonuclease subunit 2; tRNA-intron endonuclease Sen2; tRNA-intron nuclease 2; tRNA-splicing endonuclease subunit Sen2; Tsen2; TSEN2 tRNA splicing endonuclease subunit
Gene Symbols Tsen2
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-220 of human TSEN2 (NP_0795411).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 312649, 381802, 80746
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.