Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TSEN34 Mouse anti-Human, Clone: OTI6B3, lyophilized, TrueMAB™
SDP

Catalog No. 501669914
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-166-9914 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-166-9914 Supplier OriGene Supplier No. CF810146
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO

TSEN34 encodes a catalytic subunit of the tRNA splicing endonuclease, which catalyzes the removal of introns from precursor tRNAs. The endonuclease complex is also associated with a pre-mRNA 3-prime end processing factor. A mutation in this gene results in the neurological disorder pontocerebellar hypoplasia type 2. Multiple alternatively spliced variants, encoding the same protein, have been identified.

Specifications

Antigen TSEN34
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone OTI6B3
Conjugate Unconjugated
Formulation PBS with 8% trehalose and no preservative; pH 7.3
Gene TSEN34
Gene Accession No. Q9BSV6
Gene Alias LENG5, PCH2C, SEN34, SEN34L
Gene Symbols TSEN34
Host Species Mouse
Immunogen Full length human recombit protein of human TSEN34 produced in E.coli.
Purification Method Affinity Chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 79042
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Lyophilized
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.