Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TSEN34 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA559430
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA559430 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA559430 Supplier Invitrogen™ Supplier No. PA559430
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: WERGFFLSAA GKFGGDFLVY PGDPLRFHAH YIAQCWAPED TIPLQDLVAA GRLGTSVRKT LLLCSPQPDG KVVYTSLQW Highest antigen sequence identity to the following orthologs: Mouse - 95%, Rat - 95%.

TSEN34 encodes a catalytic subunit of the tRNA splicing endonuclease, which catalyzes the removal of introns from precursor tRNAs. The endonuclease complex is also associated with a pre-mRNA 3-prime end processing factor. A mutation in this gene results in the neurological disorder pontocerebellar hypoplasia type 2. Multiple alternatively spliced variants, encoding the same protein, have been identified.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TSEN34
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene TSEN34
Gene Accession No. Q9BSV6
Gene Alias CTD-3093M3.1; hsSen34; LENG5; leukocyte receptor cluster (LRC) member 5; leukocyte receptor cluster member 5; PCH2C; SEN34; SEN34L; tRNA splicing endonuclease subunit 34; tRNA-intron endonuclease Sen34; tRNA-splicing endonuclease subunit Sen34; TSEN34; TSEN34 tRNA splicing endonuclease subunit
Gene Symbols TSEN34
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human SEN34. Recombinant protein control fragment (Product #RP-99159).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 79042
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.