Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TSEN54 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PA550148
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PA550148 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PA550148 Supplier Invitrogen™ Supplier No. PA550148
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The antibody detects endogenous levels of total TSEN54 protein.

This gene encodes a subunit of the tRNA splicing endonuclease complex, which catalyzes the removal of introns from precursor tRNAs. The complex is also implicated in pre-mRNA 3-prime end processing. Mutations in this gene result in pontocerebellar hypoplasia type 2.[provided by RefSeq, Oct 2009].
TRUSTED_SUSTAINABILITY

Specifications

Antigen TSEN54
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene TSEN54
Gene Accession No. Q7Z6J9, Q8C2A2
Gene Alias 0610034P02Rik; HsSEN54; likely ortholog of H. sapiens SEN54L; PCH2A; PCH4; PCH5; sen54; SEN54 homolog; SEN54L; tRNA splicing endonuclease 54 homolog; tRNA splicing endonuclease 54 homolog (S. cerevisiae); tRNA splicing endonuclease subunit 54; tRNA-intron endonuclease Sen54; tRNA-splicing endonuclease subunit Sen54; TSEN54; TSEN54 tRNA splicing endonuclease subunit
Gene Symbols TSEN54
Host Species Rabbit
Immunogen Synthesized peptide derived from internal of human TSEN54.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 283989, 76265
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.