Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ TSG Polyclonal Antibody
Rabbit Polyclonal Antibody
$209.50 - $511.50
Specifications
Antigen | TSG |
---|---|
Concentration | 500 μg/mL |
Content And Storage | -20°C |
Applications | Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry |
Classification | Polyclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
PIPA5119492
|
Invitrogen™
PA5119492 |
200 μL |
Each for $511.50
|
|
|||||
PIPA5121522
|
Invitrogen™
PA5121522 |
20 μL |
Each for $209.50
|
|
|||||
Description
TSG Polyclonal Antibody for Western Blot, ICC/IF, IHC (P)
TWSG1 may be involved in dorsoventral axis formation. Seems to antagonize BMP signaling by forming ternary complexes with CHRD and BMPs, thereby preventing BMPs from binding to their receptors. In addition to the anti-BMP function, also has pro-BMP activity, partly mediated by cleavage and degradation of CHRD, which releases BMPs from ternary complexes. May be an important modulator of BMP-regulated cartilage development and chondrocyte differentiation. May play a role in thymocyte development.Specifications
TSG | |
-20°C | |
Polyclonal | |
Liquid | |
IgG | |
Human | |
Twsg1 | |
1810013J15Rik; 9030422N06Rik; AW552143; D17Ertd403e; PSEC0250; Tsg; Twg; twisted gastrulation BMP signaling modulator 1; twisted gastrulation homolog 1; twisted gastrulation homolog 1 (Drosophila); twisted gastrulation protein homolog 1; Twsg1 | |
Twsg1 | |
Primary | |
Antigen affinity chromatography |
500 μg/mL | |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
Unconjugated | |
Rabbit | |
RUO | |
PBS with 50% glycerol and 0.05% ProClin 300 | |
Q9GZX9 | |
57045 | |
Recombinant protein Twisted Gastrulation Protein Homolog 1. The antigen corresponds to amino acid range 26-223 of the target protein. | |
Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title