Learn More
Thermo Scientific™ TSG Series General Purpose Chromatography Refrigerators
Description
Designed for your everyday laboratory needs
Thermo Scientific™ TSG Series general-purpose chromatography refrigerators are designed with your needs in mind. The TSG Series is ideal for the demanding requirements of daily use in research, clinical, and industrial laboratories storing non-critical products such as reagents, media, proteins, and other samples requiring a 2°to 10°C controlled environment.
• Hydrocarbon safety sensor ensures cargo and lab personnel are kept safe
• Featuring a wide range of cabinet sizes from 30 cu. ft. to 72 cu. ft.
• Whisper-quiet operation
• ENERGY STAR certified
• Temperature, door ajar*, and power failure (with battery back-up) alarms
• Green, natural refrigerants
• Adaptive defrost
• Glass (double pane) or solid door configurations
• Choice of white painted interior and exterior, or stainless steel interior and gray exterior
• Digital controller
• Rear access port
• Remote alarm contacts
• Keyed door lock(s)
• LED interior lighting
• Low heat output
• 2 years parts and labor, plus 3 year compressor warranty
*All models except TSG45CPLA and TSG45CSLA come equipped with door ajar alarms

Specifications
Specifications
Description | TSG Series general purpose chromatography refrigerator |
Capacity (English) | 45 cu. ft. |
Capacity (Metric) | 1274 L |
Temperature Range | 2°C to 10°C |
Temperature (Setpoint) | +5°C |
Refrigerant | R290 |
Door Style | Sliding Glass |
No. of Doors | 2 |
Shelves | 8 Full, 2 Half |
Capacity (English) Shelf | 100 lb./full shelf; 50 lb./half shelf |
Show More |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.