Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ TSG Series General Purpose Laboratory Refrigerators

Catalog No. TSG12RPGA
Click to view available options
Capacity (Metric):
339 L
651 L
764 L
1274 L
1387 L
2038 L
Door Style:
Glass
Sliding Glass
Solid
Cabinet Material:
Gray-painted
White-painted
Attention:This item requires special delivery information. You will be asked for this during the checkout process.
Attention:This item requires special delivery information. You will be asked for this during the checkout process.

Designed for your everyday laboratory needs

Thermo Scientific™ TSG Series general-purpose laboratory refrigerators are designed with your needs in mind. The TSG Series is ideal for the demanding requirements of daily use in research, clinical, and industrial laboratories storing non-critical products such as reagents, media, proteins, and other samples requiring a 2° to 10°C controlled environment.

  • Featuring a wide range of cabinet sizes from 12 cu. ft. to 72 cu. ft.
  • Whisper-quiet operation
  • ENERGY STAR certifiedTemperature, door ajar*, and power failure (with battery back-up) alarms
  • Green, natural refrigerants
  • Adaptive defrost
  • Glass (double pane) or solid door configurations
  • Choice of white painted interior and exterior, or stainless steel interior and gray exterior
  • Digital controller
  • Rear access port
  • Remote alarm contacts
  • Keyed door lock(s)
  • LED interior lighting
  • Low heat output
  • Warranty: varies by country, please contact us for details
    *All models except TSG45RPLA and TSG45RSLA come equipped with door ajar alarm
TRUSTED_SUSTAINABILITY

Specifications

Description TSG Series general purpose laboratory refrigerator
Capacity (English) 12 cu. ft.
Capacity (Metric) 339 L
Temperature Range 2°C to 10°C
Temperature (Setpoint) +5°C
Refrigerant R290
Door Style Glass
No. of Doors 1
Shelves 4 Full
Capacity (English) Shelf 100 lb.
Capacity (Metric) Shelf 45 kg
Cabinet Material White-painted
Exterior Finish Painted
Interior Stainless Steel
Certifications/Compliance cULus, ENERGYSTAR, Prop 65
Control Type Digital Microprocessor
Ports 1 Rear access port
Warranty 2 years parts and labor, plus 3-year compressor warranty
Defrost Adaptive Defrost
Monitoring Options Alarm
Access Security Key Lock
Dimensions (D x W x H) Interior 20.75x21x45.75 in.(527x533x1162 mm)
Dimensions (L x W x H) Exterior 25x24.75x75.8 in.(635x629x1926 mm)
Amperage 5 A
Plug Type NEMA 5-15 P
Voltage 115 V
Electrical Requirements 115 V 60 Hz
Frequency 60 Hz
Green Features Energy efficient
Shipping Weight (English) 230 lb.
Shipping Weight (Metric) 105 kg
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.