Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TSG101 Antibody (JJ0900), Novus Biologicals™

Rabbit Monoclonal Antibody has been used in 2 publications
Supplier: Novus Biologicals NBP267884
Description
TSG101 Monoclonal specifically detects TSG101 in Human, Mouse, Rat samples. It is validated for Western Blot, Simple Western, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
TSG101 | |
Monoclonal | |
1 mg/mL | |
Western Blot 1:1000-1:2000, Simple Western, Flow Cytometry 1:50-1:100, Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1:100-1:500, Immunohistochemistry-Paraffin 1:50-1:200, Knockdown Validated | |
ESCRT-I complex subunit TSG101, TSG10, tumor susceptibility gene 10, tumor susceptibility gene 101, tumor susceptibility gene 101 protein, tumor susceptibility protein, VPS23 | |
Rabbit | |
Protein A purified | |
Autophagy, Cancer, Microautophagy | |
7251 | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
IgG |
Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), KnockDown | |
JJ0900 | |
Unconjugated | |
TBS (pH7.4), 0.05% BSA, 40% Glycerol with 0.05% Sodium Azide | |
TSG101 | |
Synthetic peptide within Human TSG101 aa 24-73/ 390. (SwissProt: Q99816 Human; SwissProt: Q61187 Mouse; SwissProt: Q6IRE4 Rat) | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction