Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TSG101 Monoclonal Antibody (2B7G8), CoraLite488, Proteintech
SDP

Catalog No. 502257343 Shop All Proteintech Group Inc Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-225-7343 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 50-225-7343 Supplier Proteintech Group Inc Supplier No. CL48867381100UL
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

TSG101 Monoclonal antibody specifically detects TSG101 in Human, Mouse, Rat samples. It is validated for Flow Cytometry, Immunocytochemistry, Immunohistochemistry (Paraffin)

Specifications

Antigen TSG101
Applications Flow Cytometry, Immunocytochemistry, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 2B7G8
Concentration 1 mg/mL
Conjugate CoraLite 488
Formulation PBS with 0.5% BSA, 50% glycerol and 0.05% ProClin 300; pH 7.3
Gene Hnrnph1
Gene Accession No. O35737, P31943, Q8VHV7
Gene Alias AI255943; CC2; ESCRTI complex subunit TSG101; ESCRT-I complex subunit TSG101; Rw; TSG 101; TSG10; TSG101; Tumor supressor; tumor susceptibility 101; tumor susceptibility gene 10; tumor susceptibility gene 101; tumor susceptibility gene 101 protein; tumor susceptibility protein; VPS23
Host Species Mouse
Purification Method Protein G
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 140931, 3187, 59013
Content And Storage -20° C, Avoid Freeze/Thaw Cycles, store in dark
Form Liquid
Isotype IgG1
Target Molecule Human|Mouse|Rat
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.