Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ TSG101 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA582236
Description
Immunogen sequence: RMKEEMDRAQ AELNALKRTE EDLKKGHQKL EEMVTRLDQE VAEVDKNIEL LKKKDEELSS ALEKMENQSE NNDIDEVIIP TAPLYKQILN LYAEENAIED TIFYLGEALR RGVIDLDVFL KHVRLLS.
TSG101-4A10 recognizes TSG101 protein, the product of a recently identified Tumor Susceptibility Gene whose inactivation in mouse fibroblasts results in cell transformation and the ability of those cells to form tumors in nude mice. The TSG101 gene encodes a 46 kDa protein and contains a proline-rich domain and a leucine-zipper motif within a coiled-coil domain near the carboxyl terminus.
Specifications
| TSG101 | |
| Polyclonal | |
| Unconjugated | |
| Tsg101 | |
| AI255943; CC2; ESCRTI complex subunit TSG101; ESCRT-I complex subunit TSG101; Rw; TSG 101; TSG10; TSG101; Tumor supressor; tumor susceptibility 101; tumor susceptibility gene 10; tumor susceptibility gene 101; tumor susceptibility gene 101 protein; tumor susceptibility protein; VPS23 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 22088, 292925, 7251 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.2 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q61187, Q6IRE4, Q99816 | |
| Tsg101 | |
| Recombinant protein corresponding to Human TSG101. Recombinant protein control fragment (Product #RP-89296). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction