Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TSG101 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA595851
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA595851 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA595851 Supplier Invitrogen™ Supplier No. PA595851
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: LKKKDEELSS ALEKMENQSE NNDIDEVIIP TAPLYKQILN LYAEENAIED TIFYLGEALR RGVIDLDVFL KHVRLLSRKQ FQLRALMQKA RKTAGLSDLY Positive Samples: HeLa, MCF7, K-562, Mouse spinal cord, Rat heart, Rat lung; Cellular Location: Cytoplasm, Late endosome membrane, Membrane, Nucleus, Peripheral membrane protein.

TSG101-4A10 recognizes TSG101 protein, the product of a recently identified Tumor Susceptibility Gene whose inactivation in mouse fibroblasts results in cell transformation and the ability of those cells to form tumors in nude mice. The TSG101 gene encodes a 46 kDa protein and contains a proline-rich domain and a leucine-zipper motif within a coiled-coil domain near the carboxyl terminus.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TSG101
Applications ELISA, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1.16 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.09% sodium azide; pH 7.3
Gene Tsg101
Gene Accession No. Q61187, Q6IRE4, Q99816
Gene Alias AI255943; CC2; ESCRTI complex subunit TSG101; ESCRT-I complex subunit TSG101; Rw; TSG 101; TSG10; TSG101; Tumor supressor; tumor susceptibility 101; tumor susceptibility gene 10; tumor susceptibility gene 101; tumor susceptibility gene 101 protein; tumor susceptibility protein; VPS23
Gene Symbols Tsg101
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence within amino acids 291-390 of human TSG101/VPS23 (NP_0062831).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 22088, 292925, 7251
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.