Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ TSG101 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA512040
Description
This antibody is predicted to react with mouse and rat based on sequence homology.
TSG101-4A10 recognizes TSG101 protein, the product of a recently identified Tumor Susceptibility Gene whose inactivation in mouse fibroblasts results in cell transformation and the ability of those cells to form tumors in nude mice. The TSG101 gene encodes a 46 kDa protein and contains a proline-rich domain and a leucine-zipper motif within a coiled-coil domain near the carboxyl terminus.
Specifications
TSG101 | |
Polyclonal | |
Unconjugated | |
Tsg101 | |
AI255943; CC2; ESCRTI complex subunit TSG101; ESCRT-I complex subunit TSG101; Rw; TSG 101; TSG10; TSG101; Tumor supressor; tumor susceptibility 101; tumor susceptibility gene 10; tumor susceptibility gene 101; tumor susceptibility gene 101 protein; tumor susceptibility protein; VPS23 | |
Rabbit | |
Ammonium sulfate precipitation | |
RUO | |
7251 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot | |
2.0 mg/mL | |
PBS with 0.09% sodium azide; pH 7.4 | |
Q99816 | |
Tsg101 | |
KLH conjugated synthetic peptide between 340-370 amino acids from the C-terminal region of human TSG101. | |
400 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction